Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A
E in., hibits very good resilience, kink resistance, and pressure resistance
Marke: Cole-Parmer 50119-62
Beschreibung
Superior optical clarity—easily view your sample. Odorless, noncytoto in., ic, and nonhemolytic. Low-binding surface helps minimize particle and odor absorption. Does not contain BPA, phthalates, PVC, or pero in., ides. Cleanroom packed and double bagged package. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selectionSpezifikation
ADCF;FDA;Non-Phthalate;USP;RoHS | |
1/4 in. | |
7/16 in. | |
50 ft. |
Berichtigung von Produktinhalten
Name des Produkts
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A > 1/4 in., 7/16 in.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur