missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATP1A1 (Ser16) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538261
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP dependent transport of sodium and potassium across the cell membrane. This membrane bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium / potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families.The different isoforms of the sodium / potassium ATPase exhibit tissue specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell specific expression. The alpha subunit has been found in kidney, brain, heart, and to a lesser extent liver, skeletal and smooth muscle.
Spezifikation
Phospho-ATP1A1 (Ser16) | |
Polyclonal | |
Unconjugated | |
ATP1A1 | |
Sodium/potassium-transporting ATPase subunit alpha-1, Short name=Na(+)/K(+) ATPase alpha-1 subunit, Sodium pump subunit alpha-1, ATP1A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11928, 24211, 476 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05023, Q8VDN2, P06685 | |
ATP1A1 | |
A synthetic phosphopeptide derived from human ATPase around the phosphorylation site of Ser16 (A-V-SP-E-H) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Phospho-ATP1A1 (Ser16) Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur