Learn More
SI Analytics™ pH Meter Lab 855
Modern measuring technology available along with new functionality such as AutoRead and CMC (measuring range monitoring), which makes lab measurements even more reliable. Perfectly suited for measurements in laboratories in the chemical and pharmaceutical industries as well as in medical labs.
Marke: SI Analytics™ 285206300
Beschreibung
- Reproducible measuring results with active AutoRead function
- Simple calibration with adjustable calibration timer
- Intuitive operation with clearly arranged keyboard
- Precise measurements
- Easy to operate
Spezifikation
2 to 16 | |
-10°C to +100°C | |
Lab 855 |
-2.000 bis 2.000 | |
LCD, Graphic, 128 x 64 pixel, Backlit | |
1 |