missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (PC10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Bio-Techne NB500-106C
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
PCNA | |
Monoclonal | |
DyLight 650 | |
P12004 | |
PCNA | |
Protein A-rat PCNA fusion obtained from pC2T | |
Protein A purified | |
RUO | |
Primary | |
Specific for PCNA p36 protein expressed at high levels in proliferating cells. | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PC10 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
Mouse | |
30 kDa | |
0.1 mL | |
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
5111 | |
Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish | |
Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
PCNA Antibody (PC10), DyLight 650, Novus Biologicals™ > 0.1mL; DyLight 650
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur