missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Parecoxib-Natriumsalz

Product Code. 16534484 Shop All Thermo Scientific Chemicals Products
5 g, 5 Gramm
missing translation for 'orderingAttributeHoverText'
Menge:
1 g
5 g
missing translation for 'unitSize'
1 Gramm
5 Gramm
This item is not returnable. View return policy
This item is not returnable. View return policy

Dieses Markenprodukt von Thermo Scientific Chemicals war ursprünglich Teil des Produktportfolios von Acros Organics. Auf einigen Dokumenten und Etiketteninformationen steht möglicherweise noch die bisherige Marke. Die ursprüngliche Produkt- oder Artikelnummer von Acros Organics oder die SKU-Referenz wurde bei Übernahme der Marke durch Thermo Scientific Chemicals nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198470-85-8

Specifications

Chemischer Name oder Material Parecoxib sodium salt
Schmelzpunkt 273.0°C to 275.0°C
Summenformel C19H17N2NaO4S
Menge 5 g
Molekulargewicht (g/mol) 392.4
Formelmasse 392.4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Parecoxib-Natriumsalz >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.