missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS Nylon Membrane
The intrinsic positive charge of this membrane ensures higher sensitivity and superior results even after multiple reprobing. The membrane keeps its positive charge from pH 3 to pH 12.
Marke: MP BIOMEDICALS 11MEMP0001
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The intrinsic positive charge of this membrane ensures higher sensitivity and superior results even after multiple reprobing
- Unlike some other positively charged membranes Positive™ is designed and produced specifically for blotting.
- The proprietary manufacturing method does not involve post-casting treatment steps and so results in greater lot-to-lot reproducibility.
- The membrane keeps its positive charge from pH 3 to pH 12.
Application Notes:
- Specifically for blotting. Higher sensitivity even after multiple reprobing.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
- Blotting
- Probing
- Nucleic Acid Detection

Spezifikation
3 m | |
Roll | |
3 x 0.3 m |
0.3 m | |
Nylon, Positively Charged |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
MP BIOMEDICALS Nylon Membrane > Nylon Membrane
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur