Learn More
Thermo Scientific™ Nutrient Agar (Dehydrated)
Cultivate and isolate fastidious pathogens and other micro-organisms with this nutritive, non-selective medium.
327.00 CHF
Spezifikation
Zertifizierung/Konformität | Industrienormen AFNOR, AOAC, BSI, FDA, ISO, NMKL. |
---|---|
Form | Pulver |
Produkttyp | Agar |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
10219252
|
Thermo Scientific™
CM0003B |
500 g |
327.00 CHF
500 Gramm |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
Cultivate and isolate fastidious pathogens and other microorganisms with Thermo Scientific™ Oxoid™ Nutrient Agar (Dehydrated). This non-selective medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
Nutrient Agar is a basic culture medium used to subculture organisms for maintenance purposes or to check the purity of subcultures from isolation plates prior to biochemical or serological tests.
In semi-solid form, as agar slopes or agar butts, the medium is used to maintain control organisms1.
Nutrient Agar is suitable for teaching and demonstration purposes. It contains a concentration of 1.5% agar to permit the addition of up to 10% blood or other biological fluid, as required. The medium, without additions, may be used for the cultivation of organisms which are not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Spezifikation
Industrienormen AFNOR, AOAC, BSI, FDA, ISO, NMKL. | |
Pulver | |
Agar |
For Laboratory Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.