Learn More
Novus Biologicals™ Glutathione S-Transferase pi 1/GSTP1 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke: Novus Biologicals™ NBP1-76857PEP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
A blocking peptide containing 14 amino acids near the center of human GTSP1. Source: Synthetic (Accession #: NP_000843) The Glutathione S-Transferase pi 1/GSTP1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Spezifikation
2950 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-76857. For Further Blocking Peptide Related Protocol, Click Here . | |
GSTP1 | |
0.05 mg | |
Synthetisch |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI | |
Glutathione S-Transferase pi 1/GSTP1 | |
RUO - research use only | |
14 amino acids near the center of human GTSP1 |
Berichtigung von Produktinhalten
Name des Produkts
Novus Biologicals™ Glutathione S-Transferase pi 1/GSTP1 Antibody Blocking Peptide > 0.04mg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur