missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Pinzette aus vernickeltem Stahl

Brand:  Usbeck Laborgeraete™ 3130

 View more versions of this product

Product Code. 10300802

  • 3.19 CHF / 1 Stück
Verfügbar ab: 11-06-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1/Packung
105 mm
Pinzette
Stahl; vernickelt
L 105 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Usbeck Laborgeraete™ Pinzette aus vernickeltem Stahl > Länge:105mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.