missing translation for 'onlineSavingsMsg'
Learn More
N6-Cyclopentyladenosine, Tocris Bioscience™ Artikelnummer.: 30002126
Verfügbar ab: 01-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

N6-Cyclopentyladenosine, Tocris Bioscience™

Artikelnummer. 30002126
50 mg
Click to view available options
Menge:
50 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30002126

Marke: Tocris Bioscience 1702/50

Verfügbar ab: 01-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Potent and selective A1 agonist

Potent and selective adenosine A1 receptor agonist (Ki values are 2.3, 790 and 43 nM for human A1, A2A and A3 receptors respectively; EC50 = 18600 nM for hA2B). Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Spezifikation

Chemischer Name oder Material N6-Cyclopentyladenosine
CAS 41552-82-3
Summenformel C15H21N5O4
Menge 50 mg
Synonym n6-cyclopentyladenosine, 2r,3r,4s,5r-2-6-cyclopentylamino-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, cyclopentyladenosine, n-cyclopentyladenosine, chembl68738, n 6-cyclopentyladenosine, 2r,3r,4s,5r-2-6-cyclopentylamino purin-9-yl-5-hydroxymethyl oxolane-3,4-diol, adenosine, n-cyclopentyl, n6-cyclopentylado, n-cyclopentyl-adenosine
InChI-Schlüssel SQMWSBKSHWARHU-SDBHATRESA-N
SMILES C1CCC(C1)NC2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)O)O
IUPAC-Name (2R,3R,4S,5R)-2-[6-(cyclopentylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molekulargewicht (g/mol) 335.364
PubChem CID 657378
Formelmasse 335.36
Reinheit (%) >99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N6-Cyclopentyladenosine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.