Learn More
N4BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16733-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody detects 60 kDa N4BP3 protein and a weaker 40 kDa protein.
N4BP3, also named as KIAA0341, a Fezzin family member, during nerve cell development. It is crucial for axonal and dendritic branching in developing neurons. It binds the protein NEDD4 and interacts with 14-3-3 proteins. N4BP3 is present in early neurites, growth cones, axons and dendrites of developing primary hippocampal neurons.Spezifikation
N4BP3 | |
Polyclonal | |
Unconjugated | |
N4BP3 | |
KIAA0341, N4BP3, Nedd4 binding protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
212706, 23138, 303112 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15049, Q3LUD3, Q8C7U1 | |
N4bp3 | |
N4BP3 Fusion Protein Ag10194 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Name des Produkts
N4BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur