missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-9 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marke: Gibco™ 450-30-10UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Spezifikation
P54130 | |
protein with no preservative | |
23.3 | |
10 μg | |
-20°C | |
<1 EU/μg | |
FGF9 | |
Unconjugated | |
PLGEVGSYFG VQDAVPFGNV PVLPVDSPVL LNDHLGQSEA GGLPRGPAVT DLDHLKGILR RRQLYCRTGF HLEIFPNGTI QGTRKDHSRF GILEFISIAV GLVSIRGVDS GLYLGMNEKG ELYGSEKLTQ ECVFREQFEE NWYNTYSSNL YKHVDTGRRY YVALNKDGTP REGTRTKRHQ KFTHFLPRPV DPDKVPELYK DILSQS | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
14180 | |
Mouse FGF-9 | |
E. coli | |
RUO | |
Eks; elbow knee synostosis; FGF; Fgf9; FGF-9; Fibroblast growth factor; Fibroblast growth factor 9; fibroblast growth factor 9 (glia-activating factor); GAF; glia activating factor; glia-activating factor; HBFG-9; HBGF-9; Heparin-binding growth factor 9; M-FGF-9; SYNS3 | |
Mouse | |
Recombinant | |
Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur