Learn More
Gibco™ Mouse FGF-7 (KGF) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marke: Gibco™ 450-60-500
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Spezifikation
P36363 | |
Protein with no preservative | |
18.8 | |
5 x 100 μg | |
-20°C | |
<1 EU/μg | |
FGF7 | |
Unconjugated | |
CNDMSPEQTA TSVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNSYNIMEIR TVAVGIVAIK GVESEYYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHSGG EMFVALNQKG IPVKGKKTKK EQKTAHFLPM AIT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
14178 | |
Mouse FGF-7 (KGF) | |
E. coli | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Mouse | |
Recombinant | |
Lyophilized |