Learn More
Invitrogen™ Mouse Apolipoprotein A1 ELISA Kit
ELISA
Marke:  Invitrogen™ EEL116
Artikelnummer. 17986862
  • 636.00 CHF / 1 Stück
Verfügbar ab: 29-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Q00623
0.38 ng/mL
ELISA
Colorimetric Microplate Reader
APOA1
<10
HRP
RUO
Refer to product documentation for component specific storage temperature.
1 hrs 20 mins
0.63 to 40 ng/mL
HRP
Plasma, Serum
11806
<10%
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
96 Tests
Plasma 100 μL, Serum 100 μL
ApoA1
3 hrs 30 mins
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
Berichtigung von Produktinhalten
Name des Produkts
Invitrogen™ Mouse Apolipoprotein A1 ELISA Kit > 96 Tests; HRP