missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MLK4, GST Tag Recombinant Protein
This MLK4 Recombinant Human Protein (114-420) was expressed in insect cells.
568.00 CHF - 8240.00 CHF
Spezifikation
Zugriffsnummer | NP_115811 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 60.5 kDa |
Kinase-Familie | MAP Kinases |
Proteinlänge | aa 114 - 420 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15970303
|
Invitrogen™
A30979 |
10 μg |
568.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17198745
|
Invitrogen™
A32897 |
100 μg |
2645.00 CHF
100 Mikrogramm |
Verfügbar ab: 18-11-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17168795
|
Invitrogen™
A33382 |
1 mg |
8240.00 CHF
1 Milligramm |
Verfügbar ab: 15-11-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
MLK4 or mixed lineage kinase 4 belongs to the superfamily of MAP kinase kinase kinases (MAP3K1) which contain both ser/thr and tyr kinases activity in their catalytic domains. The structure of this kinase family incorporates an N-terminal Src homology (SH3) domain, followed by the kinase domain, a leucine zipper region, and a CDC42 /RAC -interactive binding (CRIB) motif, but divergent C-terminal regions. MLK4 is highly expressed in the kidney and pancreas. MLK4 is a negative regulator of TLR4.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Spezifikation
NP_115811 | |
60.5 kDa | |
aa 114 - 420 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Recombinant, Active, Catalytic |
Kinase Assay | |
MAP Kinases | |
Human | |
GST Tag | |
Dry Ice | |
Liquid |