Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, PVDF and Viton™
Versatile check valves can be installed in any position
Marke: Cole-Parmer 50115-23
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. PVDF and Viton construction provides excellent corrosion and chemical resistance. 0.18 psi breaking pressureSpezifikation
0.1875 in. | |
3/16 in. hose barb | |
Viton |
0.12 | |
Barbed |