Learn More
Value Plastics Male Luer Slip to Female Luer Thread Adapter Fitting
Luer fittings allow for a simple yet secure connection
Marke: Value Plastics 50120-05
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standardsSpezifikation
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP | |
Male Luer Slip, Female Luer Thread | |
Polypropylene |