Learn More
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Prevent leaks with integral lock ring mechanism
26.84 CHF - 33.70 CHF
Produktcode | Marke | Material (Dichtung) | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Material (Dichtung) | Preis | Menge & Verfügbarkeit | |||||
30053948
|
Value Plastics
50120-03 |
Nylon |
26.84 CHF
25 Stück |
Verfügbar ab: 10-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
30053831
|
Value Plastics
50120-04 |
Polypropylene |
33.70 CHF
25 Stück |
Verfügbar ab: 10-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Luer fittings allow for a simple yet secure twist-lock connection. Easily transition from a male luer to female luer fitting. Ideal for general laboratory and single-use applicationsWarning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.