Learn More
MAFF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535918
Benachrichtigungen:
Um den Rabatt von 33.33% zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
Recommended positive controls: A431, A431 nuclear extract, human MAFF-transfected 293T. Predicted reactivity: Mouse (94%), Rat (96%), Chicken (80%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAFF Polyclonal antibody specifically detects MAFF in Human samples. It is validated for Western Blot.
Spezifikation
MAFF | |
Polyclonal | |
Unconjugated | |
Maff | |
hMafF; hypothetical protein MGC63868; LOW QUALITY PROTEIN: transcription factor MafF; MAF bZIP transcription factor F; Maff; Transcription factor MafF; transcription factor mafT; U-Maf; v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein F; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog f; zgc:63868 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23764 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9ULX9 | |
Maff | |
Recombinant fragment corresponding to a region within amino acids 1 and 81 of MAFF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.