Learn More
Labconco™ Logic Vue Computer Arm
Computer arm that attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse. Space-saving design eliminates need for separate computer cart.
Marke:  Labconco™ 3339000
1220.33 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Artikelnummer. 17884718
  • 1265.00 CHF / 1 Stück
Nur noch null auf Lager
Verfügbar ab: 13-10-2025
Beschreibung
  • Attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse.
  • Space-saving design eliminates need for separate computer cart.
  • VESA design supports monitors up to 18 lb.
  • Laptop Computer Arm securely supports a laptop computer within easy view of an operator.
  • Articulating arm raises, lowers and pivots to a steady position.
  • Includes articulating arm and supportive laptop platform with mounting hardware.
  • Installation is required
Spezifikation
Spezifikation
Computer Arm
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
Labconco™ Logic Vue Computer Arm > Logic Vue Computer Arm