Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
Marke:  Thermo Scientific™ N12461
 Lernen Sie weitere Varianten dieses
Artikelnummer. 30131332
  • 297.00 CHF / 1 Stück
Verfügbar ab: 17-06-2025
Beschreibung
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Nucleic Acid Extraction and Cell Separation Instrument
96 Deep-well Heating Block
Reusable
1 Block
Fresh or Frozen Plant Tissues, FFPE Tissue, Cells, Blood, Plasma
12 Samples
KingFisher Duo Prime Purification System
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
Thermo Scientific™ KingFisher Duo Prime Accessories > KingFisher Duo Prime 96 Deep-Well Heating Block