missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interferon alpha-2 Monoclonal Antibody (N27), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA135513
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MA1-35513 detects IFN-alpha II in human samples. It does not cross react with IFN-alpha I.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
Spezifikation
Interferon alpha-2 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.02% sodium azide | |
P01563 | |
IFNA2 | |
Purified recombinant IFN-alpha 2c. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
N27 | |
Unconjugated | |
IFNA2 | |
alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2 | |
Mouse | |
purified | |
RUO | |
3440 | |
4° C | |
Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Interferon alpha-2 Monoclonal Antibody (N27), Invitrogen™ > 100 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur