missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ INSTA-Blot Colon Tissue OncoPair
Applications: Western Blot
Marke: Novus Biologicals™ NBP2-30129
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Proteomic discovery tool
- Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
- Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
- Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
- Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
- Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
- Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Recommended Storage
Store at room temperature

Spezifikation
Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors | |
1 Membrane | |
PVDF |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ INSTA-Blot Colon Tissue OncoPair >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur