missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMPDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22092-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene acts as a homotetramer to regulate cell growth. The encoded protein is an enzyme that catalyzes the synthesis of xanthine monophosphate (XMP) from inosine-5′-monophosphate (IMP). This is the rate-limiting step in the de novo synthesis of guanine nucleotides. Defects in this gene are a cause of retinitis pigmentosa type 10 (RP10). Several transcript variants encoding different isoforms have been found for this gene.Spezifikation
IMPDH1 | |
Polyclonal | |
Unconjugated | |
IMPDH1 | |
IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH I, IMPDH1, LCA11, RP10, sWSS2608 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23917, 3614, 362329 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZLZ7, P20839, P50096 | |
IMPDH1 | |
IMPDH1 Fusion Protein Ag17297 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
IMPDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur