Learn More
Invitrogen™ Mouse IL-3 ELISA Kit
ELISA
950.00 CHF - 5700.00 CHF
Spezifikation
Zugriffsnummer | Gehäuse P01586 |
---|---|
Assay-Bereich | 0.16 bis 40 pg/ml |
Assay-Empfindlichkeit: | 0,2 pg/mL |
Kalibrierbereich | 0.2 bis 40 pg/ml |
Assay | Festphasen-Sandwich-ELISA |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
15415303
|
Invitrogen™
EMIL3 |
96 Tests |
950.00 CHF
1 Set |
Verfügbar ab: 02-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
17165749
|
Invitrogen™
EMIL3X5 |
5 x 96 Tests |
3140.00 CHF
1 Stück |
Verfügbar ab: 21-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
18597784
|
Invitrogen™
EMIL3X10 |
10 x 96 Tests |
5700.00 CHF
10 Stück |
Verfügbar ab: 22-07-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
The Mouse Interleukin-3 (Ms IL-3) ELISA quantitates Ms IL-3 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-3. Principle of the method The Mouse IL-3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.Spezifikation
Gehäuse P01586 | |
0,2 pg/mL | |
Festphasen-Sandwich-ELISA | |
ELISA-Kit | |
Plasma, Serum, Überstand | |
Maus | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
BPA,Csfmu,HCGF,Il-3,MCGF,PSF | |
< 10 % | |
HRP | |
Immunologie | |
2 bis 8 °C | |
Maus | |
4 h 45 min |
0.16 bis 40 pg/ml | |
0.2 bis 40 pg/ml | |
Biotin | |
Zytokine und Rezeptoren | |
Dieses ELISA-Kit zeigt keine Kreuzreaktivität mit den folgenden getesteten Zytokinen: Maus CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fraktalkin, GCSF, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-4, IL-5, IL-6, IL-9, IL-10, IL-13, KC, Leptin R, LEPTIN (OB), LIX, L-Selektin, Lymphotactin, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selektin, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TPO, VEGF. | |
ELISA, Protein-Assays, Proteinbiologie, Krebsbiologie, Immunologie, Neurowissenschaften, Neurobiologie, Proteinnachweis, Proteinanalyse | |
16187 | |
< 12 % | |
Vorbeschichtete 96-Well-Platte, Standard, Biotinylierter Nachweisantikörper, Biotinylierter Nachweisantikörper, SAV-HRP, Waschpuffer, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten | |
RUO | |
Plasma, 50 μl; Serum, 50 μl; Überstand, 100 μl | |
RP23-309E16.5, BPA, Csfmu, HCGF, Il-3, MCGF, PSF | |
1 h 20 min |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.