missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZIC3 Full-length ORF (NP_003404.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007547-P01.25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs. [provided by RefSeq]
Sequence: MTMLLDGGPQFPGLGVGSFGAPRHHEMPNREPAGMGLNPFGDSTHAAAAAAAAAAFKLSPAAAHDLSSGQSSAFTPQGSGYANALGHHHHHHHHHHHTSQVPSYGGAASAAFNSTREFLFRQRSSGLSEAASGGGQHGLFAGSASSLHAPAGIPEPPSYLLFPGLHEQGAGHPSPTGHVDNNQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFSTMHELVTHVTMEHVGGPEQNNHVCYWEECPREGKSFKAKYKLVNHIRVHTGEKPFPCPFPGCGKIFARSENLKIHKRTHTGEKPFKCEFEGCDRRFANSSDRKKHMHVHTSDKPYICKVCDKSYTHPSSLRKHMKVHESQGSDSSPAASSGYESSTPPAIASANSKDTTKTPSAVQTSTSHNPGLPPNFNEWYVSpezifikation
NP_003404.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HTX/HTX1/ZNF203 | |
ZIC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7547 | |
ZIC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTMLLDGGPQFPGLGVGSFGAPRHHEMPNREPAGMGLNPFGDSTHAAAAAAAAAAFKLSPAAAHDLSSGQSSAFTPQGSGYANALGHHHHHHHHHHHTSQVPSYGGAASAAFNSTREFLFRQRSSGLSEAASGGGQHGLFAGSASSLHAPAGIPEPPSYLLFPGLHEQGAGHPSPTGHVDNNQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFSTMHELVTHVTMEHVGGPEQNNHVCYWEECPREGKSFKAKYKLVNHIRVHTGEKPFPCPFPGCGKIFARSENLKIHKRTHTGEKPFKCEFEGCDRRFANSSDRKKHMHVHTSDKPYICKVCDKSYTHPSSLRKHMKVHESQGSDSSPAASSGYESSTPPAIASANSKDTTKTPSAVQTSTSHNPGLPPNFNEWYV | |
RUO | |
ZIC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |