Learn More
Gibco™ Human/Mouse FGF-8b Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marke: Gibco™ 100-25-5UG
83.00 CHF gültig bis 2024-12-31
Benutzen Sie den Promotions-Code "21651" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.Spezifikation
P37237, P55075 | |
protein with no preservative | |
22.5 | |
5 μg | |
-20°C | |
<1 EU/μg | |
Human, Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
14179, 2253 | |
Human/Mouse FGF-8 b | |
E. coli | |
RUO | |
FGF8 | |
Unconjugated | |
MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |