Learn More
Gibco™ Human FGF-20 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marke:  Gibco™ 100-41-250UG
 Lernen Sie weitere Varianten dieses
Artikelnummer. 17800373
  • 1524.00 CHF / 250 Mikrogramm
Verfügbar ab: 23-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Q9NP95
protein with no preservative
23.2
250 μg
-20°C
<1 EU/μg
FGF20
Unconjugated
PLAEVGGFLG GLEGLGQQVG SHFLLPPAGE RPPLLGERRS AAERSARGGP GAAQLAHLHG ILRRRQLYCR TGFHLQILPD GSVQGTRQDH SLFGILEFIS VAVGLVSIRG VDSGLYLGMN DKGELYGSEK LTSECIFREQ FEENWYNTYS SNIYKHGDTG RRYFVALNKD GTPRDGARSK RHQKFTHFLP RPVDPERVPE LYKDLLMYT
≥ 90% by SDS-PAGE gel and HPLC analyses.
Functional Assay
26281
Human FGF-20
E. coli
RUO
FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2
Human
Recombinant
Lyophilized