missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Clusterin Quantikine ELISA Kit
Human Clusterin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
1180.00 CHF - 6720.00 CHF
Spezifikation
Assay-Bereich | 3.12 - 200ng/mL |
---|---|
Assay-Empfindlichkeit: | 1.05ng/mL |
Konjugat | HRP |
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Produkttyp | Solid Phase Sandwich ELISA |
Produktcode | Marke | Menge | Ausreichend für | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Ausreichend für | Preis | Menge & Verfügbarkeit | ||||
18782354
|
R&D Systems™
DCLU00 |
1 Platte | Sufficient materials to run ELISAs on at least one 96-well plate |
1180.00 CHF
1 Set |
Verfügbar ab: 06-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
18752204
|
R&D Systems™
SCLU00 |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
6720.00 CHF
1 Set |
Verfügbar ab: 10-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
Beschreibung
The Human Clusterin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Clusterin/APOJ in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (25 ul), urine (13 ul).Spezifikation
3.12 - 200ng/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human Clusterin | |
ELISA | |
Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (25uL),Urine (13uL) | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLI, CLIclusterin (complement lysis inhibitor, SP-40, CLU, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2, TRPM-2 TRPM2 |
1.05ng/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Speichel, Serum, Urin | |
Human | |
1191 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |