missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Clusterin Quantikine ELISA Kit

Human Clusterin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Marke:  R&D Systems™ SCLU00

 Lernen Sie weitere Varianten dieses

Artikelnummer. 18752204

  • 6720.00 CHF / 1 Set
Verfügbar ab: 10-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

The Human Clusterin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Clusterin/APOJ in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (25 ul), urine (13 ul).
Spezifikation

Spezifikation

3.12 - 200ng/mL
HRP
Solid Phase Sandwich ELISA
Natural and Recombinant Human Clusterin
Human
1191
Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (25uL),Urine (13uL)
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLI, CLIclusterin (complement lysis inhibitor, SP-40, CLU, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2, TRPM-2 TRPM2
1.05ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Cell Culture Supernatant, Plasma (EDTA, Heparin), Speichel, Serum, Urin
Sufficient materials to run ELISAs on at least six 96-well plates
ELISA
6 Plates
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote