missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CFTR C-Terminus Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 42 publications
Marke: R&D Systems MAB25031
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CFTR Monoclonal specifically detects CFTR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Spezifikation
CFTR | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1080 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
24-1 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1-2 ug/10^6 cells | |
P13569 | |
CFTR | |
Glutathione S-transferase-coupled CFTR aa 1377-1480 Accession # P13569 | |
50 μg | |
Primary | |
Detects in vitro synthesized CFTR and endogenous CFTR in Western blots. | |
Human | |
Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Human CFTR C-Terminus Antibody, R&D Systems™ > 50ug; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur