Learn More
Novus Biologicals™ Human CCL1/I-309/TCA-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marke: Novus Biologicals™ NBP2-75063
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human CCL1/I-309/TCA-3 ELISA Kit (Colorimetric) measures human CCL1/I-309/TCA-3 in serum, plasma and other biological fluids.
Spezifikation
31.25 - 2000 pg/mL | |
This kit recognizes Human CCL1 in samples. No significant cross-reactivity or interference between Human CCL1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
6346 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
18.75 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CCL1 | |
1 Kit | |
CCL1/I-309/TCA-3 |
Berichtigung von Produktinhalten
Name des Produkts
Novus Biologicals™ Human CCL1/I-309/TCA-3 ELISA Kit (Colorimetric) > Quantity: 1 Kit; Host Species: Human
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur