Learn More
Heidolph™ Hei-TORQUE Ultimate 400
The Hei-TORQUE Precision line are Heidolph high performance overhead stirrers.
Marke: Heidolph™ 501-64020-04
2251.55 CHF gültig bis 2025-06-30
Benutzen Sie den Promotions-Code "24075" um Ihren Promotions-Preis zu erhalten.
Beschreibung
- The Hei-TORQUE Precision 400 laboratory stirrer is ideal for the powerful stirring of viscosities up to 250,000 mPas or volumes up to 100 liters (H2O). Thanks to its 2-gear stage design, it stirs particularly powerfully in the speed range up to 400 rpm and is therefore particularly suitable for applications with highly viscous media that require mixing in the lower speed range. However, operation at speeds of up to 2000 rpm is also possible in the second speed.
- In addition, the Hei-TORQUE Precision 400 offers, above all, the possibility of ramp programming, process monitoring (including timer function, torque trend display as a value), as well as the necessary equipment for documenting your application parameters. For this it is equipped with two interfaces - USB and RS232 - which give you the greatest flexibility to connect the device to a PC, a programmable logic controller (PLC) or an automation system such as e.g. LabManager. The scope of delivery includes the free Hei-Control software for remote control of up to 4 devices via PC.
- The laboratory stirrer Hei-TORQUE Value 400 performs every stirring task with minimal noise thanks to the latest engine technology. The safety-related start/stop touch-function serves to minimize the risk of unintentional switching on.
- Suitable for use in fume hoods or as a drive for reactor super structures, the Hei-TORQUE Precision 400 has a quick-action chuck (quick chuck with safety ring) with a diameter of 10.5 mm, which can be combined with a large number of stirring tools for the best stirring results.
Spezifikation
230 V | |
CH-Style |
Aktionen
-
Mehrere Hersteller
Die ultimative Laborgeräte-Promotion
Sparen Sie bis zu 78 % bei hochwertigen Laborgeräten und rüsten Sie Ihr Labor zu den günstigsten Preisen auf.Aktionscode: 24075Gültig bis zum: 2025-06-30
Tage
26Stunden
13Minuten
34Sekunden
32Neues Angebot Sparen Sie bis zu 78%