Learn More
Thermo Scientific™ Harvestainer™ Microcarrier Separation System
Artikelnummer. 17899786 Alle ansehen Thermo Scientific Produkte
3 l, 1 Stück
Click to view available options
Kapazität (metrisch):
3 l
12 l
Packungsgröße:
1 Stück
Artikelnummer. 17899786
Marke: Thermo Scientific™ SH31268.01
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Das Thermo Scientific Harvestainer Microcarrier Separation System ist die ideale Lösung zur Separierung der Microcarrier-Beads bei der Herstellung adhärenter Zellkulturen.
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
• Closed system
• Scalable for both smaller-scale and larger-scale applications
• Helps deliver high recovery rates
• Inline microcarrier separation
Applications
• Adherent cell culture production
TRUSTED_SUSTAINABILITY
Spezifikation
Kapazität (metrisch) 3 l
Menge 3 L
Tubing Style TPE
Berichtigung von Produktinhalten
Name des Produkts
Thermo Scientific™ Harvestainer™ Microcarrier Separation System >