Learn More
Tocris Bioscience™ GSK 1838705
Potent IR and IGF1R inhibitor; also inhibits anaplastic lymphoma kinase (ALK)
Marke:  Tocris Bioscience™ 5111/50
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18791368
  • 1085.00 CHF / 50 Milligramm
Verfügbar ab: 29-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
GSK 1838705 is a potent insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF1R) inhibitor (IC50 values are 1.6 and 2 nM, respectively). Also inhibits anaplastic lymphoma kinase (ALK) (IC50 = 0.5 nM). Displays > 800-fold selectivity for IR, IGFR1 and ALK over a panel of 44 kinases including JNK. Blocks proliferation of cancer cell lines in vitro, and causes complete regression of ALK-dependent tumors in vivo. Orally bioavailable.
Spezifikation
Spezifikation
>98%
GSK 1838705
GSK 1838705
50 mg
532.57
Nur für Forschungszwecke