missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GNAQ (Human) Recombinant Protein
Human GNAQ full-length ORF ( AAH57777, 1 a.a. - 359 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002776-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MTLESIMACCLSEEAKEARRINDEIERQLRRDKRDARRELKLLLLGTGESGKSTFIKQMRIIHGSGYSDEDKRGFTKLVYQNIFTAMQAMIRAMDTLKIPYKYEHNKAHAQLVREVDVEKVSAFENPYVDAIKSLWNDPGIQECYDRRREYQLSDSTKYYLNDLDRVADPAYLPTQQDVLRVRVPTTGIIEYPFDLQSVIFRMVDVGGQRSERRKWIHCFENVTSIMFLVALSEYDQVLVESDNENRMEESKALFRTIITYPWFQNSSVILFLNKKDLLEEKIMYSHLVDYFPEYDGPQRDAQAAREFILKMFVDLNPDSDKIIYSHFTCATDTENIRFVFAAVKDTILQLNLKEYNLV
Specifications
AAH57777 | |
guanine nucleotide binding protein (G protein), q polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNAQ | |
GST |
2776 | |
Wheat germ expression system | |
25 μg | |
G-ALPHA-q, GAQ | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |