Learn More
Proteintech FlexAble FITC Plus Antibody Labeling Kit for Human IgG
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier.
Marke:  Proteintech KFA109-200
Additional Details : Gewicht : 0.02000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18030596
  • 1009.87 CHF / 200 Reaktionen
Verfügbar ab: 05-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
Spezifikation
100 Tests