Learn More
Proteintech FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier.
Marke:  Proteintech KFA105-200
Additional Details : Gewicht : 0.02000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18084735
  • 1009.87 CHF / 200 Reaktionen
Verfügbar ab: 05-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
FlexAble CoraLite™ Plus 555 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 555 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
Spezifikation
100 Tests