Learn More
Proteintech FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier.
102.24 CHF - 1009.87 CHF
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18078705
|
Proteintech
KFA104-50 |
50 Tests |
475.97 CHF
50 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18089415
|
Proteintech
KFA104-200 |
100 Tests |
1009.87 CHF
200 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18024235
|
Proteintech
KFA104-10 |
10 Tests |
102.24 CHF
10 Reaktionen |
Verfügbar ab: 03-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
FlexAble CoraLite™ Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.