Learn More
Proteintech FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier.
Marke:  Proteintech KFA104-10
Additional Details : Gewicht : 0.02000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18024235
  • 102.24 CHF / 10 Reaktionen
Verfügbar ab: 05-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
FlexAble CoraLite™ Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
Spezifikation
10 Tests