missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10060-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.Spezifikation
FKBPL | |
Polyclonal | |
Unconjugated | |
FKBPL | |
DIR1, FK506 binding protein like, FKBPL, NG7, WISP39 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
406168, 56299, 63943 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35450, Q6MG81, Q9UIM3 | |
Fkbpl | |
FKBPL Fusion Protein Ag0112 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
FKBPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur