Learn More
EYA1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519220
456.40 CHF gültig bis 2025-03-16
Benutzen Sie den Promotions-Code "24070" um Ihren Promotions-Preis zu erhalten.
Benachrichtigungen:
Um den Rabatt von 33.33% zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
Spezifikation
EYA1 | |
Polyclonal | |
Unconjugated | |
EYA1 | |
BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2138 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q99502 | |
EYA1 | |
Synthetic peptide sequence (TDPTAEYSTIHSP) corresponding to the internal amino acids of EYA1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.