missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC6/CSB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24291-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.Spezifikation
ERCC6/CSB | |
Polyclonal | |
Unconjugated | |
ERCC6 | |
ARMD5, ATP dependent helicase ERCC6, CKN2, Cockayne syndrome protein CSB, COFS, COFS1, CSB, ERCC6, RAD26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2074 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q03468 | |
ERCC6 | |
ERCC6/CSB Fusion Protein Ag18009 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
ERCC6/CSB Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur