missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Equine IFN-gamma ELISA Kit

ELISA

906.00 CHF - 3140.00 CHF

Spezifikation

Zugriffsnummer BoIFNG; Zytokin; EGK_03901; gamma-Interferon; gamma-Interferon-Vorläufer; gamma-Interferon; gamma-Interferon-Vorläufer (AA -23 bis 143); H-IFN-g; Ifg; IFI; IFN-gamma; IFN ?; IFNG; IFN-g; IFNG2; IFN-gamma; IFN-gamma-Vorläufer; IFN-y; IFN?; Immun-Interferon; Interferon gamma; Interferon gamma-Vorläufer; Interferon gamma Typ 2; Interferon y; Interferon ?; Interferon, gamma; Interferon-gamma; Interferon-gamma; Interferon-gamma-Pegel; Interferon-gamma-Vorläufer; Interferon?; M-IFN-g; R-IFN-g
Assay-Bereich 0,13 bis 100 ng/ml
Assay-Empfindlichkeit: 0,13 ng/mL
Konjugat Biotin
Produkttyp ELISA
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17174819
Unterlagen ansehen
Invitrogen™
EE1RBX5
5 x 96 Tests
3140.00 CHF
1 Stück
Nur noch null auf Lager
Verfügbar ab: 21-07-2025
Zum Warenkorb hinzufügen
 
17184353
Unterlagen ansehen
Invitrogen™
EE1RB
96 Tests
906.00 CHF
96 Stück
Nur noch null auf Lager
Verfügbar ab: 22-07-2025
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Equine IFN-gamma quantitates equine IFN-gamma in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant equine IFN-gamma.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Spezifikation
Mehr anzeigen
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Invitrogen™ Equine IFN-gamma ELISA Kit

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt