missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (C225 (Cetuximab)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Marke: Bio-Techne NBP2-75903-0.2MG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize.
Spezifikation
EGFR | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
EGFR | |
The epidermal growth factor receptor is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. | |
Primary | |
Recognises human EGFR. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Flow Cytometry, Immunohistochemistry, Blocking Assay, Neutralization | |
C225 (Cetuximab) | |
Flow Cytometry, Immunohistochemistry, Block/Neutralize | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Human | |
0.2 mg | |
1956 | |
Human | |
IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
EGFR Antibody (C225 (Cetuximab)) - Azide and BSA Free, Novus Biologicals™ > EGFR Antibody; Unconjugated; 0.2 mg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur