missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin / CD324 Mouse, Clone: 4A2, enQuireBio™
Mouse Monoclonal Antibody
234.00 CHF - 484.00 CHF
Spezifikation
Antigen | E-Cadherin / CD324 |
---|---|
Klon | 4A2 |
Konzentration | 200μg/mL |
Anwendungen | Immunohistochemistry |
Klassifikation | Monoclonal |
Spezifikation
E-Cadherin / CD324 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
999 | |
Recombinant human full-length E-Cadherin protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
120-80kDa (Mature) ;135kDa (Precursor) |
4A2 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001304114 | |
CDH1 | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface | |
LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. |