missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin / CD324 Mouse, Clone: 4A2, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 999-MSM4-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
E-Cadherin / CD324 | |
Cell Surface | |
4A2 | |
Unconjugated | |
NP_001304114 | |
Mouse | |
120-80kDa (Mature) ;135kDa (Precursor) | |
Protein A/G Purified | |
RUO | |
999 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CDH1 | |
Recombinant human full-length E-Cadherin protein | |
LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |