Learn More
Invitrogen™ Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488
Donkey Polyclonal Secondary Antibody
Marke: Invitrogen™ SA510086
Beschreibung
This antibody is cross-adsorbed and exhibits minimum reactivity to chicken, human, mouse, pig, rabbit and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
Goat IgG (H+L) Cross-Adsorbed, Goat IgG (H+L) | |
Polyclonal | |
DyLight 488 | |
Donkey | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Secondary Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 6.8 to 7.4 | |
Goat IgG-heavy and light chain. | |
500 μg | |
Secondary | |
4°C | |
Liquid |
Zertifikate
Eine Chargennummer ist erforderlich, um Ergebnisse für Zertifikate anzuzeigen. Um Ihre Chargennummer zu früheren Bestellungen zu finden, können Sie unsere Auftragsverfolgung nutzen.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.