missing translation for 'onlineSavingsMsg'
Learn More
DNQX, Tocris Bioscience™ Artikelnummer.: 30001059
Verfügbar ab: 01-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

DNQX, Tocris Bioscience™

Product Code. 30001059
50 mg
missing translation for 'orderingAttributeHoverText'
Menge:
10 mg
50 mg
This item is not returnable. View return policy

Artikelnummer. 30001059

Marke: Tocris Bioscience 0189/50

Verfügbar ab: 01-04-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
This item is not returnable. View return policy

Selective non-NMDA iGluR antagonist

Selective non-NMDA receptor antagonist.

Water-soluble Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2379-57-9
Summenformel C8H4N4O6
Molekulargewicht (g/mol) 252.14
MDL-Nummer MFCD00069257
InChI-Schlüssel RWVIMCIPOAXUDG-UHFFFAOYSA-N
Synonym dnqx, unii-62t278s1mx, 6,7-dinitroquinoxaline-2,3-dione dnqx, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dinitro, 6,7-dinitroquinoxaline-2,3-diol, 6,7-dinitro-1,4-dihydro-quinoxaline-2,3-dione, 6,7-dinitroquinoxaline-2,3 1h,4h-dione, 1ftl, tocris-0189, lopac-d-0540
PubChem CID 3899541
IUPAC-Name 6,7-dinitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione
SMILES [O-][N+](=O)C1=C(C=C2NC(=O)C(=O)NC2=C1)[N+]([O-])=O

Specifications

Chemischer Name oder Material DNQX
Menge 50 mg
Formelmasse 252.14
Reinheit (%) >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DNQX, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.