Learn More
Cytochrome B5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538934
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene is a membrane-bound cytochrome that reduces ferric hemoglobin (methemoglobin) to ferrous hemoglobin, which is required for stearyl-CoA-desaturase activity. Defects in this gene are a cause of type IV hereditary methemoglobinemia. Three transcript variants encoding different isoforms have been found for this gene.Spezifikation
Cytochrome b5 | |
Polyclonal | |
Unconjugated | |
CYB5A | |
CYB5, MCB5, cytochrome b5, type 1 cyt-b5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109672, 1528, 64001 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P00167, P56395, P00173 | |
CYB5A | |
A synthetic peptide derived from the internal region of human CYB5 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Name des Produkts
Cytochrome B5 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur